missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Bromobenzyl)-4-methylperhydro-1,4-diazepine, 97%, Maybridge

Chemical Identifiers

CAS 280560-78-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C13H19BrN2
Formula Weight 283.21
MDL Number MFCD00810550
Chemical Name or Material 1-(4-Bromobenzyl)-4-methylperhydro-1, 4-diazepine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC55810DE SDS Fisher Scientific Company
CC55810DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC55810DA SDS Fisher Scientific Company
CC55810DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

280560-78-3
100.0
C13H19BrN2
MFCD00810550
97%
96.5
97%
283.21
1-(4-Bromobenzyl)-4-methylperhydro-1, 4-diazepine

Specifications

283.21
100.0
C13H19BrN2
Amber Glass Bottle
97%
97%
5g
MFCD00810550
96.5
280560-78-3
1-(4-Bromobenzyl)-4-methylperhydro-1, 4-diazepine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger