missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Bromo-2-fluorobenzyl)piperazine, 96%

$72.65 - $933.78

Chemical Identifiers

CAS 870703-75-6
Molecular Formula C11H14BrFN2
Molecular Weight (g/mol) 273.149
MDL Number MFCD07368744
InChI Key NRCBEINBLSTDHM-UHFFFAOYSA-N
Synonym 1-4-bromo-2-fluorobenzyl piperazine
PubChem CID 2782804
IUPAC Name 1-[(4-bromo-2-fluorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=C(C=C(C=C2)Br)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5584403 View Documents Alfa Aesar™
H5584403
1g Each for $72.65
 
AAH5584406 View Documents Alfa Aesar™
H5584406
5g Each for $276.43
 
AAH5584414 View Documents Alfa Aesar™
H5584414
25g Each for $933.78
 
Specifications

Chemical Identifiers

870703-75-6
273.149
NRCBEINBLSTDHM-UHFFFAOYSA-N
2782804
C1CN(CCN1)CC2=C(C=C(C=C2)Br)F
C11H14BrFN2
MFCD07368744
1-4-bromo-2-fluorobenzyl piperazine
1-[(4-bromo-2-fluorophenyl)methyl]piperazine

Specifications

1.411 g/mL
1.563
Yellow to Orange
870703-75-6
MFCD07368744
NRCBEINBLSTDHM-UHFFFAOYSA-N
1-[(4-bromo-2-fluorophenyl)methyl]piperazine
2782804
Liquid
1-(4-Bromo-2-fluorobenzyl)piperazine
>110°C (230°F)
120°C to 122°C (0.02mmHg)
1g
C11H14BrFN2
1-4-bromo-2-fluorobenzyl piperazine
C1CN(CCN1)CC2=C(C=C(C=C2)Br)F
273.149
273.15
96%
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures