missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Bromo-2-fluorobenzyl)piperazine, 96%

Manufacturer:  Alfa Aesar™ H5584414

 View more versions of this product

Catalog No. AAH5584414



Specifications

Specifications

1-(4-Bromo-2-fluorobenzyl)piperazine
>110°C (230°F)
120°C to 122°C (0.02mmHg)
25g
C11H14BrFN2
1-4-bromo-2-fluorobenzyl piperazine
C1CN(CCN1)CC2=C(C=C(C=C2)Br)F
273.149
273.15
96%
1.411 g/mL
1.563
Yellow to Orange
870703-75-6
MFCD07368744
NRCBEINBLSTDHM-UHFFFAOYSA-N
1-[(4-bromo-2-fluorophenyl)methyl]piperazine
2782804
Liquid

Chemical Identifiers

870703-75-6
273.149
NRCBEINBLSTDHM-UHFFFAOYSA-N
2782804
C1CN(CCN1)CC2=C(C=C(C=C2)Br)F
C11H14BrFN2
MFCD07368744
1-4-bromo-2-fluorobenzyl piperazine
1-[(4-bromo-2-fluorophenyl)methyl]piperazine
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents