missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(4-Boc-1-piperidinyl)-4-bromopyrazole, 98%, Thermo Scientific Chemicals
$140.78 - $499.40
Chemical Identifiers
CAS | 877399-50-3 |
---|---|
Molecular Formula | C13H20BrN3O2 |
Molecular Weight (g/mol) | 330.23 |
MDL Number | MFCD09831980 |
InChI Key | IYNZAVDBHAQODX-UHFFFAOYSA-N |
Synonym | tert-butyl 4-4-bromo-1h-pyrazol-1-yl piperidine-1-carboxylate, 1-boc-4-4-bromopyrazol-1-yl piperidine, 4-4-bromopyrazol-1-yl piperidine-1-carboxylic acid tert-butyl ester, 4-4-bromo-pyrazol-1-yl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-bromopyrazol-1-yl piperidine-1-carboxylate, n-boc-4-4-bromopyrazol-1-yl piperidine, 3-2-ethoxycarbonylacetyl piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-bromo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester, ksc658s5d, 1-boc-4-4-bromo-1-pyrazolyl piperidine |
PubChem CID | 45480278 |
IUPAC Name | tert-butyl 4-(4-bromopyrazol-1-yl)piperidine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCC(CC1)N1C=C(Br)C=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6531503
|
Thermo Scientific Chemicals
H6531503 |
1 g |
N/A
|
|
|||||
AAH6531506
|
Thermo Scientific Chemicals
H6531506 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
877399-50-3 | |
330.23 | |
IYNZAVDBHAQODX-UHFFFAOYSA-N | |
45480278 | |
CC(C)(C)OC(=O)N1CCC(CC1)N1C=C(Br)C=N1 |
C13H20BrN3O2 | |
MFCD09831980 | |
tert-butyl 4-4-bromo-1h-pyrazol-1-yl piperidine-1-carboxylate, 1-boc-4-4-bromopyrazol-1-yl piperidine, 4-4-bromopyrazol-1-yl piperidine-1-carboxylic acid tert-butyl ester, 4-4-bromo-pyrazol-1-yl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-bromopyrazol-1-yl piperidine-1-carboxylate, n-boc-4-4-bromopyrazol-1-yl piperidine, 3-2-ethoxycarbonylacetyl piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-bromo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester, ksc658s5d, 1-boc-4-4-bromo-1-pyrazolyl piperidine | |
tert-butyl 4-(4-bromopyrazol-1-yl)piperidine-1-carboxylate |
Specifications
877399-50-3 | |
C13H20BrN3O2 | |
1 g | |
tert-butyl 4-4-bromo-1h-pyrazol-1-yl piperidine-1-carboxylate, 1-boc-4-4-bromopyrazol-1-yl piperidine, 4-4-bromopyrazol-1-yl piperidine-1-carboxylic acid tert-butyl ester, 4-4-bromo-pyrazol-1-yl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-bromopyrazol-1-yl piperidine-1-carboxylate, n-boc-4-4-bromopyrazol-1-yl piperidine, 3-2-ethoxycarbonylacetyl piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-bromo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester, ksc658s5d, 1-boc-4-4-bromo-1-pyrazolyl piperidine | |
CC(C)(C)OC(=O)N1CCC(CC1)N1C=C(Br)C=N1 | |
330.23 | |
330.23 | |
Solid |
White | |
MFCD09831980 | |
14511017 | |
IYNZAVDBHAQODX-UHFFFAOYSA-N | |
tert-butyl 4-(4-bromopyrazol-1-yl)piperidine-1-carboxylate | |
45480278 | |
98% | |
1-(4-Boc-1-piperidinyl)-4-bromopyrazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only