missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Boc-1-piperidinyl)-4-bromopyrazole, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6531506

 View more versions of this product

Catalog No. AAH6531506



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-(4-Boc-1-piperidinyl)-4-bromopyrazole
White
MFCD09831980
14511017
IYNZAVDBHAQODX-UHFFFAOYSA-N
tert-butyl 4-(4-bromopyrazol-1-yl)piperidine-1-carboxylate
45480278
98%
877399-50-3
C13H20BrN3O2
5 g
tert-butyl 4-4-bromo-1h-pyrazol-1-yl piperidine-1-carboxylate, 1-boc-4-4-bromopyrazol-1-yl piperidine, 4-4-bromopyrazol-1-yl piperidine-1-carboxylic acid tert-butyl ester, 4-4-bromo-pyrazol-1-yl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-bromopyrazol-1-yl piperidine-1-carboxylate, n-boc-4-4-bromopyrazol-1-yl piperidine, 3-2-ethoxycarbonylacetyl piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-bromo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester, ksc658s5d, 1-boc-4-4-bromo-1-pyrazolyl piperidine
CC(C)(C)OC(=O)N1CCC(CC1)N1C=C(Br)C=N1
330.23
330.23
Solid

Chemical Identifiers

877399-50-3
330.23
IYNZAVDBHAQODX-UHFFFAOYSA-N
45480278
CC(C)(C)OC(=O)N1CCC(CC1)N1C=C(Br)C=N1
C13H20BrN3O2
MFCD09831980
tert-butyl 4-4-bromo-1h-pyrazol-1-yl piperidine-1-carboxylate, 1-boc-4-4-bromopyrazol-1-yl piperidine, 4-4-bromopyrazol-1-yl piperidine-1-carboxylic acid tert-butyl ester, 4-4-bromo-pyrazol-1-yl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-bromopyrazol-1-yl piperidine-1-carboxylate, n-boc-4-4-bromopyrazol-1-yl piperidine, 3-2-ethoxycarbonylacetyl piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-bromo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester, ksc658s5d, 1-boc-4-4-bromo-1-pyrazolyl piperidine
tert-butyl 4-(4-bromopyrazol-1-yl)piperidine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only