Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Bis(trimethylsilyl)tetrafluorobenzene, 99%

$795.57 - $795.57

Chemical Identifiers

Chemical Name or Material 1,4-Bis(trimethylsilyl)tetrafluorobenzene
CAS 16956-91-5
Molecular Formula C12H18F4Si2
Formula Weight 294.44
MDL Number MFCD00093109
Synonym Tetrafluoro-1,4-bis(trimethylsilyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2600903

  SDS
alfa aesar™
H2600903
1g Each for $795.57
Description & Specifications

Specifications

Formula Weight 294.44
Chemical Name or Material 1,4-Bis(trimethylsilyl)tetrafluorobenzene
Quantity 1g
Synonym Tetrafluoro-1,4-bis(trimethylsilyl)benzene
CAS 16956-91-5
Melting Point 53° to 55°C
MDL Number MFCD00093109
Assay 99%
Molecular Formula C12H18F4Si2