Please login to your online account to display your discounted pricing

1,4-Bis[(trimethylsilyl)ethynyl]benzene 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC390620010

 View more versions of this product

Catalog No. AC390620010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 270.52
Quantity 1g
Color White
Melting Point 119° to 123°C
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 1, 4-Bis[(trimethylsilyl)ethynyl]benzene
Assay Percent Range 98%
MDL Number MFCD00135433
CAS Min % 97.5
Packaging Glass bottle
Linear Formula C6H4[CCSi(CH3)3]2
Molecular Formula C16H22Si2
CAS 17938-13-5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1, 4-Bis[(trimethylsilyl)ethynyl]benzene
CAS 17938-13-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H22Si2
Linear Formula C6H4[CCSi(CH3)3]2
Formula Weight 270.52
MDL Number MFCD00135433
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents