Please login to your online account to display your discounted pricing

1,4-Bis[(trimethylsilyl)ethynyl]benzene 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 4-Bis[(trimethylsilyl)ethynyl]benzene
CAS 17938-13-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C16H22Si2
Linear Formula C6H4[CCSi(CH3)3]2
Formula Weight 270.52
MDL Number MFCD00135433
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390620010

  SDS
thermo fisher scientific
AC390620010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 270.52
Quantity 1g
Color White
Melting Point 119° to 123°C
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 1, 4-Bis[(trimethylsilyl)ethynyl]benzene
Packaging Glass bottle
Assay Percent Range 98%
MDL Number MFCD00135433
CAS Min % 97.5
Molecular Formula C16H22Si2
Linear Formula C6H4[CCSi(CH3)3]2
CAS 17938-13-5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.