Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Bis[(trimethylsilyl)ethynyl]benzene, 98%

$38.80 - $38.80

Chemical Identifiers

Chemical Name or Material 1,4-Bis[(trimethylsilyl)ethynyl]benzene
CAS 17938-13-5
Molecular Formula C16H22Si2
Formula Weight 270.53
MDL Number MFCD00135433
Beilstein 2840618
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0970903

  SDS
alfa aesar™
L0970903
1g Each for $38.80
Description & Specifications

Specifications

Formula Weight 270.53
Chemical Name or Material 1,4-Bis[(trimethylsilyl)ethynyl]benzene
Molecular Formula C16H22Si2
CAS 17938-13-5
Beilstein 2840618
Melting Point 119° to 121°C
Assay 98%
MDL Number MFCD00135433
Quantity 1g