Please login to your online account to display your discounted pricing

1,4-Bis(trichloromethyl)benzene 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  154170100

 View more versions of this product

Catalog No. AC154170100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder and Chunks
Formula Weight 312.83
Quantity 10g
Color Light yellow
EINECS Number 200-686-3
TSCA TSCA
Melting Point 107° to 112°C
RTECS Number ZE4655000
Beilstein 05, 385
Merck Index 15, 1304
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 1, 4-Bis(trichloromethyl)benzene
Molecular Formula C8H4Cl6
CAS 68-36-0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula C6H4(CCl3)2
MDL Number MFCD00000791
CAS Min % 98.5
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Synonym α,α,α,α',α',α'-Hexachloro-p-xylene
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material 1, 4-Bis(trichloromethyl)benzene
Name Note 99%
CAS 68-36-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H4Cl6
Linear Formula C6H4(CCl3)2
Formula Weight 312.83
MDL Number MFCD00000791
Synonym α,α,α,α',α',α'-Hexachloro-p-xylene
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents