Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Bis(trans-2-phenylethenyl)benzene, 97%

$322.45 - $322.45

Chemical Identifiers

Chemical Name or Material 1,4-Bis(trans-2-phenylethenyl)benzene
CAS 1608-41-9
Molecular Formula C22H18
Formula Weight 282.39
MDL Number MFCD00154950
Synonym 1,4-Di(trans-beta-styryl)benzene; trans-4-beta-Styryl-trans stilbene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3143603

  SDS
alfa aesar™
H3143603
1g Each for $322.45
Description & Specifications

Specifications

Formula Weight 282.39
Chemical Name or Material 1,4-Bis(trans-2-phenylethenyl)benzene
Assay 97%
Molecular Formula C22H18
MDL Number MFCD00154950
CAS 1608-41-9
Synonym 1,4-Di(trans-beta-styryl)benzene; trans-4-beta-Styryl-trans stilbene
Melting Point 264° to 266°C
Quantity 1g