missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Bis(methylamino)anthraquinone, 95%

$57.05 - $183.51

Chemical Identifiers

CAS 2475-44-7
Molecular Formula C16H14N2O2
Molecular Weight (g/mol) 266.3
MDL Number MFCD00001198
InChI Key QOSTVEDABRQTSU-UHFFFAOYSA-N
Synonym disperse blue 14, 1,4-bis methylamino anthraquinone, resiren blue tb, solvent blue 78, acetate blue b, celliton fast blue b, cibacet blue br, setacyl blue bs, teraprint blue r, 1,4-bis methylamino anthracene-9,10-dione
PubChem CID 17189
IUPAC Name 1,4-bis(methylamino)anthracene-9,10-dione
SMILES CNC1=C2C(=C(C=C1)NC)C(=O)C3=CC=CC=C3C2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0090306 View Documents Alfa Aesar™
L0090306
5g Each for $57.05
 
AAL0090314 View Documents Alfa Aesar™
L0090314
25g Each for $183.51
 
Specifications

Chemical Identifiers

2475-44-7
266.3
QOSTVEDABRQTSU-UHFFFAOYSA-N
17189
CNC1=C2C(=C(C=C1)NC)C(=O)C3=CC=CC=C3C2=O
C16H14N2O2
MFCD00001198
disperse blue 14, 1,4-bis methylamino anthraquinone, resiren blue tb, solvent blue 78, acetate blue b, celliton fast blue b, cibacet blue br, setacyl blue bs, teraprint blue r, 1,4-bis methylamino anthracene-9,10-dione
1,4-bis(methylamino)anthracene-9,10-dione

Specifications

~215°C
2475-44-7
MFCD00001198
disperse blue 14, 1,4-bis methylamino anthraquinone, resiren blue tb, solvent blue 78, acetate blue b, celliton fast blue b, cibacet blue br, setacyl blue bs, teraprint blue r, 1,4-bis methylamino anthracene-9,10-dione
CNC1=C2C(=C(C=C1)NC)C(=O)C3=CC=CC=C3C2=O
266.3
266.3
1,4-Bis(methylamino)anthraquinone
5g
C16H14N2O2
2220693
QOSTVEDABRQTSU-UHFFFAOYSA-N
1,4-bis(methylamino)anthracene-9,10-dione
17189
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CB5750000

TSCA : TSCA