missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Bis(hexafluoro-alpha-hydroxyisopropyl)benzene Hydrate, TCI America™

$140.56 - $484.31

Chemical Identifiers

CAS 1992-15-0
Molecular Formula C12H6F12O2
Molecular Weight (g/mol) 410.159
MDL Number MFCD00192487
InChI Key YTJDSANDEZLYOU-UHFFFAOYSA-N
Synonym 1,4-bis hexafluoro-alpha-hydroxyisopropyl benzene, 2,2'-1,4-phenylene bis 1,1,1,3,3,3-hexafluoropropan-2-ol, 1,4-bis 2-hydroxyhexafluoroisopropyl benzene, 1,4-bis perfluoro 2-hydroxyprop-2-yl benzene, 1,1,1,3,3,3-hexafluoro-2-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,4-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,4-benzenedimethanol, 1,1,1,3,3,3-hexafluoro-2-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethy l phenyl propan-2-ol, 2,2'-p-phenylene bis 1,1,1,3,3,3-hexafluoro-2-propanol, 1,4-bis 2-hydroxyhexafluoro-2-propyl benzene
PubChem CID 631065
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES C1=CC(=CC=C1C(C(F)(F)F)(C(F)(F)F)O)C(C(F)(F)F)(C(F)(F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B14195G View Documents TCI America
B14195G
5g Each for $140.56
 
B141925G View Documents TCI America
B141925G
25g Each for $484.31
 
Specifications

Chemical Identifiers

1992-15-0
410.159
YTJDSANDEZLYOU-UHFFFAOYSA-N
631065
C1=CC(=CC=C1C(C(F)(F)F)(C(F)(F)F)O)C(C(F)(F)F)(C(F)(F)F)O
C12H6F12O2
MFCD00192487
1,4-bis hexafluoro-alpha-hydroxyisopropyl benzene, 2,2'-1,4-phenylene bis 1,1,1,3,3,3-hexafluoropropan-2-ol, 1,4-bis 2-hydroxyhexafluoroisopropyl benzene, 1,4-bis perfluoro 2-hydroxyprop-2-yl benzene, 1,1,1,3,3,3-hexafluoro-2-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,4-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,4-benzenedimethanol, 1,1,1,3,3,3-hexafluoro-2-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethy l phenyl propan-2-ol, 2,2'-p-phenylene bis 1,1,1,3,3,3-hexafluoro-2-propanol, 1,4-bis 2-hydroxyhexafluoro-2-propyl benzene
1,1,1,3,3,3-hexafluoro-2-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol

Specifications

White
5g
C12H6F12O2
1,4-bis hexafluoro-alpha-hydroxyisopropyl benzene, 2,2'-1,4-phenylene bis 1,1,1,3,3,3-hexafluoropropan-2-ol, 1,4-bis 2-hydroxyhexafluoroisopropyl benzene, 1,4-bis perfluoro 2-hydroxyprop-2-yl benzene, 1,1,1,3,3,3-hexafluoro-2-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,4-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,4-benzenedimethanol, 1,1,1,3,3,3-hexafluoro-2-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethy l phenyl propan-2-ol, 2,2'-p-phenylene bis 1,1,1,3,3,3-hexafluoro-2-propanol, 1,4-bis 2-hydroxyhexafluoro-2-propyl benzene
C1=CC(=CC=C1C(C(F)(F)F)(C(F)(F)F)O)C(C(F)(F)F)(C(F)(F)F)O
410.159
410.16
≥98.0% (GC)
94°C
1992-15-0
MFCD00192487
YTJDSANDEZLYOU-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoro-2-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
631065
Crystal-Powder at 20°C
1,4-Bis(hexafluoro-alpha-hydroxyisopropyl)benzene Hydrate
Safety and Handling

Safety and Handling

TSCA : No