Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Bis(glycidyloxy)benzene, 97%

$586.62 - $586.62

Chemical Identifiers

Chemical Name or Material 1,4-Bis(glycidyloxy)benzene
CAS 2425-01-6
Molecular Formula C12H14O4
Formula Weight 222.24
MDL Number MFCD00186891
Synonym Hydroquinone diglycidyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2062814

 
alfa aesar™
B2062814
25g Each for $586.62
Description & Specifications

Specifications

Formula Weight 222.24
Chemical Name or Material 1,4-Bis(glycidyloxy)benzene
Synonym Hydroquinone diglycidyl ether
Melting Point 110° to 112°C
TSCA TSCA
Assay 97%
Molecular Formula C12H14O4
MDL Number MFCD00186891
CAS 2425-01-6
Health Hazard 1 Warning
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
is a good general-purpose inhibitor, stabilizer, antioxidant, and intermediate. It is used as a photographic reducer and developer.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.