Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Bis(diphenylphosphino)butane, 98%

$22.74 - $253.14

Chemical Identifiers

Chemical Name or Material 1,4-Bis(diphenylphosphino)butane
CAS 7688-25-7
Molecular Formula C28H28P2
Linear Formula (C6H5)2P(CH2)4P(C6H5)2
Formula Weight 426.48
MDL Number MFCD00003051
Beilstein 706446
Synonym Dppb
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2112214

  SDS
alfa aesar™
B2112214
25g Each for $253.14

AAB2112203

  SDS
alfa aesar™
B2112203
1g Each for $22.74

AAB2112206

  SDS
alfa aesar™
B2112206
5g Each for $61.64
Description & Specifications

Specifications

Formula Weight 426.48
Linear Formula (C6H5)2P(CH2)4P(C6H5)2
Quantity 25g
Assay 98%
Synonym Dppb
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 706446
Melting Point 132° to 136°C
Health Hazard 1 Warning
Molecular Formula C28H28P2
Chemical Name or Material 1,4-Bis(diphenylphosphino)butane
MDL Number MFCD00003051
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
CAS 7688-25-7

Applications
1,4-Bis(diphenylphosphino)butane is used in Suzuki reaction. It plays an essential role as a ligand in palladium(0)-catalyzed acylcyanation of arylacetylenes and deprotection of allyloxycarbamates. It acts as a ligand for alkylations, isomerization reactions and in organometallic chemistry. Further, it is used in Heck reaction, Negishi coupling and Sonogashira coupling. In addition to this, it is useful in organometallic chemistry.

Solubility
Soluble in chloroform. Slightly soluble in water

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.