Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Bis(dimethylsilyl)benzene, 97%

$124.92 - $441.49

Chemical Identifiers

Chemical Name or Material 1,4-Bis (dimethylsilyl)benzene
CAS 2488-01-9
Molecular Formula C10H18Si2
Formula Weight 194.43
MDL Number MFCD00039790
Beilstein 1072658
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1212606

  SDS
alfa aesar™
A1212606
5g Each for $124.92

AAA1212614

  SDS
alfa aesar™
A1212614
25g Each for $441.49
Description & Specifications

Specifications

Formula Weight 194.43
Boiling Point 213° to 214°C
Health Hazard 1 Warning
Quantity 5g
Chemical Name or Material 1,4-Bis (dimethylsilyl)benzene
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Health Hazard 2 GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Molecular Formula C10H18Si2
TSCA TSCA
CAS 2488-01-9
Beilstein 1072658
Flash Point 77°C (170°F)
MDL Number MFCD00039790
RTECS Number VV4830000
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.