missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(aminomethyl)cyclohexane (cis- and trans- mixture) 98.0+%, TCI America™

$139.81 - $337.40

Chemical Identifiers

CAS 2549-93-1
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00039463
InChI Key OXIKYYJDTWKERT-UHFFFAOYSA-N
Synonym 1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanedimethanamine, bamch, cis-1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanebis methylamine, cyclohexane-1,4-diyldimethanamine, trans-1,4-bis aminomethyl cyclohexane, 4-aminomethyl cyclohexyl methanamine, trans-1,4-cyclohexanedimethanamine, cyclohex-1,4-ylenebis methylamine
PubChem CID 17354
IUPAC Name [4-(aminomethyl)cyclohexyl]methanamine
SMILES C1CC(CCC1CN)CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B10835G
View Documents
TCI America
B10835G
5 g
Each for $139.81
Only null left
 
B108325G
View Documents
TCI America
B108325G
25 g
Each for $337.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2549-93-1
142.246
OXIKYYJDTWKERT-UHFFFAOYSA-N
17354
C1CC(CCC1CN)CN
C8H18N2
MFCD00039463
1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanedimethanamine, bamch, cis-1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanebis methylamine, cyclohexane-1,4-diyldimethanamine, trans-1,4-bis aminomethyl cyclohexane, 4-aminomethyl cyclohexyl methanamine, trans-1,4-cyclohexanedimethanamine, cyclohex-1,4-ylenebis methylamine
[4-(aminomethyl)cyclohexyl]methanamine

Specifications

2549-93-1
C8H18N2
5 g
1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanedimethanamine, bamch, cis-1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanebis methylamine, cyclohexane-1,4-diyldimethanamine, trans-1,4-bis aminomethyl cyclohexane, 4-aminomethyl cyclohexyl methanamine, trans-1,4-cyclohexanedimethanamine, cyclohex-1,4-ylenebis methylamine
C1CC(CCC1CN)CN
142.246
142.25
Liquid
Yellow
MFCD00039463
2922
OXIKYYJDTWKERT-UHFFFAOYSA-N
[4-(aminomethyl)cyclohexyl]methanamine
17354
≥98.0% (GC,T)
1,4-Bis(aminomethyl)cyclohexane (cis- and trans- mixture)
Videos
Safety and Handling

Safety and Handling

RTECSNumber : GU7000010

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'