missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Bis(aminomethyl)cyclohexane (cis- and trans- mixture) 98.0+%, TCI America™
Supplier: TCI America B108325G
Specifications
| 1,4-Bis(aminomethyl)cyclohexane (cis- and trans- mixture) | |
| Yellow | |
| MFCD00039463 | |
| 2922 | |
| OXIKYYJDTWKERT-UHFFFAOYSA-N | |
| [4-(aminomethyl)cyclohexyl]methanamine | |
| 17354 | |
| ≥98.0% (GC,T) |
| 2549-93-1 | |
| C8H18N2 | |
| 25 g | |
| 1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanedimethanamine, bamch, cis-1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanebis methylamine, cyclohexane-1,4-diyldimethanamine, trans-1,4-bis aminomethyl cyclohexane, 4-aminomethyl cyclohexyl methanamine, trans-1,4-cyclohexanedimethanamine, cyclohex-1,4-ylenebis methylamine | |
| C1CC(CCC1CN)CN | |
| 142.246 | |
| 142.25 | |
| Liquid |
Chemical Identifiers
| 2549-93-1 | |
| 142.246 | |
| OXIKYYJDTWKERT-UHFFFAOYSA-N | |
| 17354 | |
| C1CC(CCC1CN)CN |
| C8H18N2 | |
| MFCD00039463 | |
| 1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanedimethanamine, bamch, cis-1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanebis methylamine, cyclohexane-1,4-diyldimethanamine, trans-1,4-bis aminomethyl cyclohexane, 4-aminomethyl cyclohexyl methanamine, trans-1,4-cyclohexanedimethanamine, cyclohex-1,4-ylenebis methylamine | |
| [4-(aminomethyl)cyclohexyl]methanamine |
Safety and Handling
RTECSNumber : GU7000010
TSCA : Yes