Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Bis(5-phenyloxazol-2-yl)benzene, 98+%

$72.01 - $272.73

Chemical Identifiers

Chemical Name or Material 1,4-Bis (5-phenyloxazol-2-yl)benzene
CAS 1806-34-4
Molecular Formula C24H16N2O2
Formula Weight 364.41
MDL Number MFCD00005309
Beilstein 325741
Synonym POPOP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1232106

  SDS
alfa aesar™
A1232106
5g Each for $72.01

AAA1232114

  SDS
alfa aesar™
A1232114
25g Each for $272.73
Description & Specifications

Specifications

Formula Weight 364.41
Chemical Name or Material 1,4-Bis (5-phenyloxazol-2-yl)benzene
TSCA TSCA
Assay ≥98%
Molecular Formula C24H16N2O2
MDL Number MFCD00005309
CAS 1806-34-4
Beilstein 325741
Synonym POPOP
Melting Point 241° to 243°C
Quantity 5g