missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Bis(2-propynyloxy)benzene, TCI America™

$149.72 - $516.41

Chemical Identifiers

CAS 34596-36-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00224313
InChI Key ODCQCPUUPWRQKV-UHFFFAOYSA-N
Synonym 1,4-Bis(propargyloxy)benzene
PubChem CID 12933255
IUPAC Name 1,4-bis(prop-2-ynoxy)benzene
SMILES C#CCOC1=CC=C(C=C1)OCC#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B46071G View Documents TCI America
B46071G
1g Each for $149.72
 
B46075G View Documents TCI America
B46075G
5g Each for $516.41
 
Specifications

Chemical Identifiers

34596-36-6
186.21
ODCQCPUUPWRQKV-UHFFFAOYSA-N
12933255
C#CCOC1=CC=C(C=C1)OCC#C
C12H10O2
MFCD00224313
1,4-Bis(propargyloxy)benzene
1,4-bis(prop-2-ynoxy)benzene

Specifications

202°C
50°C
34596-36-6
MFCD00224313
ODCQCPUUPWRQKV-UHFFFAOYSA-N
1,4-bis(prop-2-ynoxy)benzene
12933255
Crystal-Powder at 20°C
1,4-Bis(2-propynyloxy)benzene
White-Yellow
1g
C12H10O2
1,4-Bis(propargyloxy)benzene
C#CCOC1=CC=C(C=C1)OCC#C
186.21
186.21
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator