Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Bis(2-methylstyryl)benzene, 99%

$52.49 - $187.80

Chemical Identifiers

Chemical Name or Material 1,4-Bis(2-methylstyryl)benzene
CAS 13280-61-0
Molecular Formula C24H22
Formula Weight 310.44
MDL Number MFCD00008529
Beilstein 2375845
Synonym Bis-MSB; 1,4-Di(2-methylstyryl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1353406

 
alfa aesar™
A1353406
5g Each for $52.49

AAA1353414

 
alfa aesar™
A1353414
25g Each for $187.80
Description & Specifications

Specifications

Formula Weight 310.44
Chemical Name or Material 1,4-Bis(2-methylstyryl)benzene
TSCA TSCA
Assay 99%
Molecular Formula C24H22
MDL Number MFCD00008529
CAS 13280-61-0
Beilstein 2375845
Synonym Bis-MSB; 1,4-Di(2-methylstyryl)benzene
Melting Point 179° to 184°C
Quantity 5g
Health Hazard 1 Warning
Odor Odorless
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.