missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2025909
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene | |
92°C to 95°C | |
C12H6F12O2 | |
10 g | |
YTJDSANDEZLYOU-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexafluoro-2-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol | |
631065 | |
98% |
1992-15-0 | |
100°C (20 mmHg) | |
MFCD00042091 | |
1,4-bis hexafluoro-alpha-hydroxyisopropyl benzene, 2,2'-1,4-phenylene bis 1,1,1,3,3,3-hexafluoropropan-2-ol, 1,4-bis 2-hydroxyhexafluoroisopropyl benzene, 1,4-bis perfluoro 2-hydroxyprop-2-yl benzene, 1,1,1,3,3,3-hexafluoro-2-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,4-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,4-benzenedimethanol, 1,1,1,3,3,3-hexafluoro-2-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethy l phenyl propan-2-ol, 2,2'-p-phenylene bis 1,1,1,3,3,3-hexafluoro-2-propanol, 1,4-bis 2-hydroxyhexafluoro-2-propyl benzene | |
OC(C1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F)(C(F)(F)F)C(F)(F)F | |
410.16 | |
410.16 |
Chemical Identifiers
1992-15-0 | |
410.16 | |
YTJDSANDEZLYOU-UHFFFAOYSA-N | |
631065 | |
OC(C1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F)(C(F)(F)F)C(F)(F)F |
C12H6F12O2 | |
MFCD00042091 | |
1,4-bis hexafluoro-alpha-hydroxyisopropyl benzene, 2,2'-1,4-phenylene bis 1,1,1,3,3,3-hexafluoropropan-2-ol, 1,4-bis 2-hydroxyhexafluoroisopropyl benzene, 1,4-bis perfluoro 2-hydroxyprop-2-yl benzene, 1,1,1,3,3,3-hexafluoro-2-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,4-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,4-benzenedimethanol, 1,1,1,3,3,3-hexafluoro-2-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethy l phenyl propan-2-ol, 2,2'-p-phenylene bis 1,1,1,3,3,3-hexafluoro-2-propanol, 1,4-bis 2-hydroxyhexafluoro-2-propyl benzene | |
1,1,1,3,3,3-hexafluoro-2-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only