missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Bis(2-hydroxyethyl)piperazine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5506314
Description
It is applied for desulfurization, decarburization extraction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is applied for desulfurization, decarburization extraction.
Solubility
Soluble in water.
Notes
Store at 4°C. Keep away from oxidizing agents.
Specifications
1,4-Bis(2-hydroxyethyl)piperazine | |
133°C to 136°C | |
C8H18N2O2 | |
25 g | |
Soluble in water. | |
C1CN(CCN1CCO)CCO | |
174.244 | |
174.24 |
122-96-3 | |
215°C to 220°C (50 mmHg) | |
MFCD00006157 | |
1,4-piperazinediethanol, 2,2'-piperazine-1,4-diyl diethanol, 1,4-bis 2-hydroxyethyl piperazine, n,n'-bis 2-hydroxyethyl piperazine, 1,4-di 2-hydroxyethyl piperazine, n,n'-dihydroxyethylpiperazine, n,n'-di 2-hydroxyethyl piperazine, unii-eq349pkx8y, piperazine, n,n'-bis 2-hydroxyethyl, 1,4-bis beta-hydroxyethyl piperazine | |
VARKIGWTYBUWNT-UHFFFAOYSA-N | |
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanol | |
67151 | |
99% |
Chemical Identifiers
122-96-3 | |
174.244 | |
VARKIGWTYBUWNT-UHFFFAOYSA-N | |
67151 | |
C1CN(CCN1CCO)CCO |
C8H18N2O2 | |
MFCD00006157 | |
1,4-piperazinediethanol, 2,2'-piperazine-1,4-diyl diethanol, 1,4-bis 2-hydroxyethyl piperazine, n,n'-bis 2-hydroxyethyl piperazine, 1,4-di 2-hydroxyethyl piperazine, n,n'-dihydroxyethylpiperazine, n,n'-di 2-hydroxyethyl piperazine, unii-eq349pkx8y, piperazine, n,n'-bis 2-hydroxyethyl, 1,4-bis beta-hydroxyethyl piperazine | |
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanol |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 204-586-0
RTECSNumber : TL3675000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only