missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Bis(2-hydroxyethyl)benzene, TCI America™

$433.89 - $1,671.42

Chemical Identifiers

CAS 5140-03-4
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00666548
InChI Key LBZZJNPUANNABV-UHFFFAOYSA-N
Synonym 1,4-Benzenediethanol, 1,4-Phenylenediethanol
PubChem CID 5013205
IUPAC Name 2-[4-(2-hydroxyethyl)phenyl]ethanol
SMILES C1=CC(=CC=C1CCO)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B22955G View Documents TCI America
B22955G
5g Each for $433.89
 
B229525G View Documents TCI America
B229525G
25g Each for $1,671.42
 
Specifications

Chemical Identifiers

5140-03-4
166.22
LBZZJNPUANNABV-UHFFFAOYSA-N
5013205
C1=CC(=CC=C1CCO)CCO
C10H14O2
MFCD00666548
1,4-Benzenediethanol, 1,4-Phenylenediethanol
2-[4-(2-hydroxyethyl)phenyl]ethanol

Specifications

193°C
88°C
5140-03-4
MFCD00666548
LBZZJNPUANNABV-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)phenyl]ethanol
5013205
Crystal-Powder at 20°C
1,4-Bis(2-hydroxyethyl)benzene
White
5g
C10H14O2
1,4-Benzenediethanol, 1,4-Phenylenediethanol
C1=CC(=CC=C1CCO)CCO
166.22
166.22
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No