Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Bis(2-hydroxy-3,5-di-tert-butylbenzyl)piperazine, 95%

$60.88 - $244.09

Chemical Identifiers

Chemical Name or Material 1,4-Bis(2-hydroxy-3,5-di-tert-butylbenzyl)piperazine
CAS 110546-20-8
Molecular Formula C34H54N2O2
Formula Weight 522.81
MDL Number MFCD14156144
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5190403

  SDS
alfa aesar™
H5190403
1g Each for $60.88

AAH5190406

  SDS
alfa aesar™
H5190406
5g Each for $244.09
Description & Specifications

Specifications

Formula Weight 522.81
Quantity 1g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
MDL Number MFCD14156144
Molecular Formula C34H54N2O2
Chemical Name or Material 1,4-Bis(2-hydroxy-3,5-di-tert-butylbenzyl)piperazine
Assay 95%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 110546-20-8
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.