missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Bis[2-(4-methyl-5-phenyloxazolyl)]benzene [for scintillation spectrometry], TCI America™

$154.31 - $592.80

Chemical Identifiers

CAS 3073-87-8
Molecular Formula C26H20N2O2
Molecular Weight (g/mol) 392.458
MDL Number MFCD00005310
InChI Key VLDFXDUAENINOO-UHFFFAOYSA-N
Synonym dimethyl-popop, 1,4-bis 4-methyl-5-phenyloxazol-2-yl benzene, oxazole, 2,2'-1,4-phenylene bis 4-methyl-5-phenyl, dimethyl popop, 1,4-bis-2-4-methyl-5-phenyloxazoylyl benzene, oxazole, 2,2'-p-phenylenebis 4-methyl-5-phenyl, 4-methyl-2-4-4-methyl-5-phenyl-1,3-oxazol-2-yl phenyl-5-phenyl-1,3-oxazole, 1,4-bis 4-methyl-5-phenyl-2-oxazolyl benzene, oxazole, 2,2'-1,4-phenylene bis 4-methyl-5-phenyl, 1,4-bis 4-methyl-5-phenyloxazol-2-yl benzene dimethyl popop
PubChem CID 76491
IUPAC Name 4-methyl-2-[4-(4-methyl-5-phenyl-1,3-oxazol-2-yl)phenyl]-5-phenyl-1,3-oxazole
SMILES CC1=C(OC(=N1)C2=CC=C(C=C2)C3=NC(=C(O3)C4=CC=CC=C4)C)C5=CC=CC=C5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B04995G View Documents TCI America
B04995G
5g Each for $154.31
 
B049925G View Documents TCI America
B049925G
25g Each for $592.80
 
Specifications

Chemical Identifiers

3073-87-8
392.458
VLDFXDUAENINOO-UHFFFAOYSA-N
76491
CC1=C(OC(=N1)C2=CC=C(C=C2)C3=NC(=C(O3)C4=CC=CC=C4)C)C5=CC=CC=C5
C26H20N2O2
MFCD00005310
dimethyl-popop, 1,4-bis 4-methyl-5-phenyloxazol-2-yl benzene, oxazole, 2,2'-1,4-phenylene bis 4-methyl-5-phenyl, dimethyl popop, 1,4-bis-2-4-methyl-5-phenyloxazoylyl benzene, oxazole, 2,2'-p-phenylenebis 4-methyl-5-phenyl, 4-methyl-2-4-4-methyl-5-phenyl-1,3-oxazol-2-yl phenyl-5-phenyl-1,3-oxazole, 1,4-bis 4-methyl-5-phenyl-2-oxazolyl benzene, oxazole, 2,2'-1,4-phenylene bis 4-methyl-5-phenyl, 1,4-bis 4-methyl-5-phenyloxazol-2-yl benzene dimethyl popop
4-methyl-2-[4-(4-methyl-5-phenyl-1,3-oxazol-2-yl)phenyl]-5-phenyl-1,3-oxazole

Specifications

Undesignated
5g
C26H20N2O2
dimethyl-popop, 1,4-bis 4-methyl-5-phenyloxazol-2-yl benzene, oxazole, 2,2'-1,4-phenylene bis 4-methyl-5-phenyl, dimethyl popop, 1,4-bis-2-4-methyl-5-phenyloxazoylyl benzene, oxazole, 2,2'-p-phenylenebis 4-methyl-5-phenyl, 4-methyl-2-4-4-methyl-5-phenyl-1,3-oxazol-2-yl phenyl-5-phenyl-1,3-oxazole, 1,4-bis 4-methyl-5-phenyl-2-oxazolyl benzene, oxazole, 2,2'-1,4-phenylene bis 4-methyl-5-phenyl, 1,4-bis 4-methyl-5-phenyloxazol-2-yl benzene dimethyl popop
CC1=C(OC(=N1)C2=CC=C(C=C2)C3=NC(=C(O3)C4=CC=CC=C4)C)C5=CC=CC=C5
392.458
392.46
≥99.0% (HPLC,N)
230°C
3073-87-8
MFCD00005310
VLDFXDUAENINOO-UHFFFAOYSA-N
4-methyl-2-[4-(4-methyl-5-phenyl-1,3-oxazol-2-yl)phenyl]-5-phenyl-1,3-oxazole
76491
Crystal-Powder at 20°C
1,4-Bis[2-(4-methyl-5-phenyloxazolyl)]benzene [for scintillation spectrometry]
Safety and Handling

Safety and Handling

TSCA : Yes