missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Bis[2-(4-methyl-5-phenyloxazolyl)]benzene 99.0+%, TCI America™

Manufacturer:  TCI America B04995G

 View more versions of this product

Catalog No. B04995G



Specifications

Specifications

1,4-Bis[2-(4-methyl-5-phenyloxazolyl)]benzene [for scintillation spectrometry]
230°C
3073-87-8
MFCD00005310
VLDFXDUAENINOO-UHFFFAOYSA-N
4-methyl-2-[4-(4-methyl-5-phenyl-1,3-oxazol-2-yl)phenyl]-5-phenyl-1,3-oxazole
76491
Crystal-Powder at 20°C
Undesignated
5g
C26H20N2O2
1,4-bis 4-methyl-5-phenyl-2-oxazolyl benzene
CC1=C(OC(=N1)C2=CC=C(C=C2)C3=NC(=C(O3)C4=CC=CC=C4)C)C5=CC=CC=C5
392.458
392.46
≥99.0% (HPLC,N)

Chemical Identifiers

3073-87-8
392.458
VLDFXDUAENINOO-UHFFFAOYSA-N
76491
CC1=C(OC(=N1)C2=CC=C(C=C2)C3=NC(=C(O3)C4=CC=CC=C4)C)C5=CC=CC=C5
C26H20N2O2
MFCD00005310
1,4-bis 4-methyl-5-phenyl-2-oxazolyl benzene
4-methyl-2-[4-(4-methyl-5-phenyl-1,3-oxazol-2-yl)phenyl]-5-phenyl-1,3-oxazole
Safety and Handling

Safety and Handling

TSCA : Yes

Documents