Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Bis(1-hydroxycyclohexyl)-1,3-butadiyne, 98+%

$280.26 - $280.26

Chemical Identifiers

Chemical Name or Material 1,4-Bis(1-hydroxycyclohexyl)-1,3-butadiyne
CAS 5768-10-5
Molecular Formula C16H22O2
Formula Weight 246.35
MDL Number MFCD00021401
Beilstein 1246867
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1249214

  SDS
alfa aesar™
L1249214
25g Each for $280.26
Description & Specifications

Specifications

Formula Weight 246.35
Chemical Name or Material 1,4-Bis(1-hydroxycyclohexyl)-1,3-butadiyne
Assay ≥98%
MDL Number MFCD00021401
CAS 5768-10-5
Beilstein 1246867
RTECS Number GV8650000
Molecular Formula C16H22O2
Melting Point 174° to 175°C
Quantity 25g