Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Biphenylyl)ethanol, 98%

$252.45 - $252.45

Chemical Identifiers

Chemical Name or Material 1-(4-Biphenylyl)ethanol
CAS 3562-73-0
Molecular Formula C14H14O
Formula Weight 198.27
MDL Number MFCD00016859
Beilstein 2044710
Synonym 4-Biphenylyl methyl carbinol; 4-(1-Hydroxyethyl)biphenyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0754309

 
alfa aesar™
L0754309
10g Each for $252.45
Description & Specifications

Specifications

Formula Weight 198.27
Chemical Name or Material 1-(4-Biphenylyl)ethanol
Assay 98%
Molecular Formula C14H14O
MDL Number MFCD00016859
CAS 3562-73-0
Beilstein 2044710
Melting Point 94° to 99°C
Synonym 4-Biphenylyl methyl carbinol; 4-(1-Hydroxyethyl)biphenyl
Quantity 10g