Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Benzodioxane-6-carboxylic acid, 95%

Chemical Identifiers

Chemical Name or Material 1,4-Benzodioxane-6-carboxylic acid
CAS 4442-54-0
Formula Weight 180.16
MDL Number MFCD00463509
Beilstein 177824
Synonym 2,3-Dihydro-1,4-benzodioxin-6-ylcarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6699206

 
alfa aesar™
H6699206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6699214

 
alfa aesar™
H6699214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.16
Melting Point 134° to 138°C
Chemical Name or Material 1,4-Benzodioxane-6-carboxylic acid
Assay 95%
MDL Number MFCD00463509
CAS 4442-54-0
Synonym 2,3-Dihydro-1,4-benzodioxin-6-ylcarboxylic acid
Quantity 5g
Physical Form Crystalline Solid
Beilstein 177824
Color Pale brown