Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Benzodioxane-2-thiocarboxamide, 97%

$67.11 - $507.96

Chemical Identifiers

Chemical Name or Material 1,4-Benzodioxane-2-thiocarboxamide
CAS 147031-85-4
Molecular Formula C9H9NO2S
Formula Weight 195.24
MDL Number MFCD00084889
Synonym 2,3-Dihydro-1,4-benzodioxin-2-carbothioamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2657103

  SDS
alfa aesar™
H2657103
1g Each for $160.08

AAH2657106

  SDS
alfa aesar™
H2657106
5g Each for $507.96

AAH26571MD

  SDS
alfa aesar™
H26571MD
250mg Each for $67.11
Description & Specifications

Specifications

Formula Weight 195.24
Chemical Name or Material 1,4-Benzodioxane-2-thiocarboxamide
Assay 97%
Molecular Formula C9H9NO2S
MDL Number MFCD00084889
CAS 147031-85-4
Synonym 2,3-Dihydro-1,4-benzodioxin-2-carbothioamide
Melting Point 172° to 176°C
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.