missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Benzodioxane-2-carboxylic acid, 97%

$69.86 - $69.86

Chemical Identifiers

CAS 3663-80-7
Molecular Formula C9H8O4
Formula Weight 180.16
MDL Number MFCD00084888
Chemical Name or Material 1,4-Benzodioxane-2-carboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2066103 SDS Alfa Aesar™
B2066103
1g Each for $69.86
AAB2066106 SDS Alfa Aesar™
B2066106
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

3663-80-7
180.16
1,4-Benzodioxane-2-carboxylic acid
C9H8O4
MFCD00084888

Specifications

3663-80-7
180.16
122° to 126°C
1,4-Benzodioxane-2-carboxylic acid
C9H8O4
MFCD00084888
1g
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning