Please login to your online account to display your discounted pricing

1,4-Benzodioxan, 99%, ACROS Organics™

$96.48 - $96.48

Chemical Identifiers

Chemical Name or Material 1, 4-Benzodioxan
CAS 493-09-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H8O2
Formula Weight 136.15
MDL Number MFCD00006821
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC176731000

  SDS
thermo fisher scientific
AC176731000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC176730250

  SDS
thermo fisher scientific
176730250
Glass bottle 25mL Each for $96.48
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥98.5%
Formula Weight 136.15
Quantity 100mL
Color Clear colorless to light yellow
Density 1.1400g/mL
Beilstein 19, 22
CAS Max % 100.0
Chemical Name or Material 1, 4-Benzodioxan
Infrared Spectrum Authentic
Assay Percent Range 99%
MDL Number MFCD00006821
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.5485 to 1.5505 (20°C, 589nm)
Molecular Formula C8H8O2
CAS 493-09-4