Please login to your online account to display your discounted pricing

1,4-Benzodioxan-5-carboxylic acid, 98%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC399610010

 View more versions of this product

Catalog No. AC399610010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 1g
EINECS Number 224-670-0
Chemical Name or Material 1, 4-Benzodioxan-5-carboxylic acid
CAS 4442-53-9
Assay Percent Range 98%
Molecular Formula C9H8O4
MDL Number MFCD00239415

Specifications

Chemical Name or Material 1, 4-Benzodioxan-5-carboxylic acid
CAS 4442-53-9
Assay Percent Range 98%
Molecular Formula C9H8O4
MDL Number MFCD00239415