Please login to your online account to display your discounted pricing

1,4-Benzodioxan-5-carboxylic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 4-Benzodioxan-5-carboxylic acid
CAS 4442-53-9
Assay Percent Range 98%
Molecular Formula C9H8O4
MDL Number MFCD00239415
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC399610010

  SDS
thermo fisher scientific
AC399610010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC399610050

  SDS
thermo fisher scientific
AC399610050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
EINECS Number 224-670-0
Chemical Name or Material 1, 4-Benzodioxan-5-carboxylic acid
Assay Percent Range 98%
Molecular Formula C9H8O4
MDL Number MFCD00239415
CAS 4442-53-9