missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Benzodioxan-5-amine, 95%, ACROS Organics™

Chemical Identifiers

CAS 16081-45-1
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C8H9NO2
Formula Weight 151.17
MDL Number MFCD03695459
Chemical Name or Material 1, 4-Benzodioxan-5-amine
Assay ≥94.0% (GC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC429370010 SDS Thermo Fisher Scientific
AC429370010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC429370050 SDS Thermo Fisher Scientific
AC429370050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

16081-45-1
100.0
C8H9NO2
MFCD03695459
≥94.0% (GC)
94.0
95%
151.17
1, 4-Benzodioxan-5-amine

Specifications

Spherical Beads
1g
94.0
Authentic
MFCD03695459
95%
1, 4-Benzodioxan-5-amine
151.17
Yellow to amber
100.0
16081-45-1
C8H9NO2
Glass bottle
≥94.0% (GC)

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning