missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Benzenedimethanol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1440522
Description
1,4-Benzenedimethanol is used as an organic synthesis intermediate. It is also used for preparation of soluble polyphenylene and p-tolyl-methanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Benzenedimethanol is used as an organic synthesis intermediate. It is also used for preparation of soluble polyphenylene and p-tolyl-methanol.
Solubility
Soluble in ether, toluene, methanol and hot water.
Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
Specifications
1,4-Benzenedimethanol | |
117°C to 121°C | |
C8H10O2 | |
100 g | |
1,4-benzenedimethanol, p-xylylene glycol, p-xylene glycol, terephthalyl alcohol, 4-hydroxymethyl phenyl methanol, 1,4-xylylene glycol, 1,4-phenylenedimethanol, p-benzenedimethanol, p-xylylenediol, 1,4-dimethylolbenzene | |
BWVAOONFBYYRHY-UHFFFAOYSA-N | |
[4-(hydroxymethyl)phenyl]methanol | |
11506 | |
99% |
589-29-7 | |
138°C to 143°C (1 mmHg) | |
MFCD00004665 | |
2042077 | |
Soluble in ether,toluene,methanol and hot water. | |
C1=CC(=CC=C1CO)CO | |
138.166 | |
138.17 |
Chemical Identifiers
589-29-7 | |
138.166 | |
BWVAOONFBYYRHY-UHFFFAOYSA-N | |
11506 | |
C1=CC(=CC=C1CO)CO |
C8H10O2 | |
MFCD00004665 | |
1,4-benzenedimethanol, p-xylylene glycol, p-xylene glycol, terephthalyl alcohol, 4-hydroxymethyl phenyl methanol, 1,4-xylylene glycol, 1,4-phenylenedimethanol, p-benzenedimethanol, p-xylylenediol, 1,4-dimethylolbenzene | |
[4-(hydroxymethyl)phenyl]methanol |
Safety and Handling
EINECSNumber : 209-641-2
TSCA : Yes
RUO – Research Use Only