Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Benzenediboronic acid bis(neopentyl glycol) ester, 99%

$97.19 - $1,014.05

Chemical Identifiers

Chemical Name or Material 1,4-Benzenediboronic acid bis(neopentyl glycol) ester
CAS 5565-36-6
Molecular Formula C16H24B2O4
Formula Weight 301.99
MDL Number MFCD01075726
Beilstein 2952607
Synonym 1,4-Benzenediboronic acid bis-2,2-dimethyltrimethylene ester; 2,2'-(1,4-Phenylene)bis(5,5-dimethyl-1,3,2-dioxaborinane)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1618706

  SDS
alfa aesar™
L1618706
5g Each for $1,014.05

AAL1618703

  SDS
alfa aesar™
L1618703
1g Each for $287.80

AAL16187MD

  SDS
alfa aesar™
L16187MD
250mg Each for $97.19
Description & Specifications

Specifications

Formula Weight 301.99
Chemical Name or Material 1,4-Benzenediboronic acid bis(neopentyl glycol) ester
MDL Number MFCD01075726
Assay 99%
Molecular Formula C16H24B2O4
Melting Point 231° to 233°C
Quantity 5g
CAS 5565-36-6
Synonym 1,4-Benzenediboronic acid bis-2,2-dimethyltrimethylene ester; 2,2'-(1,4-Phenylene)bis(5,5-dimethyl-1,3,2-dioxaborinane)
Beilstein 2952607