missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine 96.0+%, TCI America™

$148.44 - $440.45

Chemical Identifiers

CAS 74852-62-3
Molecular Formula C17H21N3O
Molecular Weight (g/mol) 283.375
MDL Number MFCD02071496
InChI Key VXEGSRKPIUDPQT-UHFFFAOYSA-N
Synonym 4-[4-(4-Methoxyphenyl)-1-piperazinyl]aniline
PubChem CID 154406
IUPAC Name 4-[4-(4-methoxyphenyl)piperazin-1-yl]aniline
SMILES COC1=CC=C(C=C1)N2CCN(CC2)C3=CC=C(C=C3)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19205G View Documents Tci America
M19205G
5g Each for $148.44
 
M192025G View Documents Tci America
M192025G
25g Each for $440.45
 
Specifications

Chemical Identifiers

74852-62-3
283.375
VXEGSRKPIUDPQT-UHFFFAOYSA-N
154406
COC1=CC=C(C=C1)N2CCN(CC2)C3=CC=C(C=C3)N
C17H21N3O
MFCD02071496
4-[4-(4-Methoxyphenyl)-1-piperazinyl]aniline
4-[4-(4-methoxyphenyl)piperazin-1-yl]aniline

Specifications

Undesignated
5g
C17H21N3O
4-[4-(4-Methoxyphenyl)-1-piperazinyl]aniline
COC1=CC=C(C=C1)N2CCN(CC2)C3=CC=C(C=C3)N
283.375
283.38
≥96.0% (HPLC,T)
186°C
74852-62-3
MFCD02071496
VXEGSRKPIUDPQT-UHFFFAOYSA-N
4-[4-(4-methoxyphenyl)piperazin-1-yl]aniline
154406
Crystal-Powder at 20°C
1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine
Safety and Handling

Safety and Handling

TSCA : No