Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Aminobenzyl)-1H-1,2,4-triazole, 98%

Chemical Identifiers

Chemical Name or Material 1-(4-Aminobenzyl)-1H-1,2,4-triazole
CAS 119192-10-8
Formula Weight 174.21
MDL Number MFCD00973306
UN Number UN2811
Beilstein 5934828
Synonym 4-(1H-1,2,4-Triazol-1-ylmethyl)aniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6662014

  SDS
alfa aesar™
H6662014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6662022

  SDS
alfa aesar™
H6662022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 126° to 130°C
Formula Weight 174.21
Physical Form Powder
Assay 98%
Health Hazard 1 Warning
Chemical Name or Material 1-(4-Aminobenzyl)-1H-1,2,4-triazole
Color White to pale yellow
Quantity 25g
MDL Number MFCD00973306
CAS 119192-10-8
UN Number UN2811
Beilstein 5934828
Synonym 4-(1H-1,2,4-Triazol-1-ylmethyl)aniline
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.