missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Amino-2-n-propyl-5-pyrimidinylmethyl)-2-methylpyridinium chloride

A thiamine analogue; blocks thiamine uptake and prevents carbohydrate synthesis

$103.95 - $374.73

Chemical Identifiers

CAS 137-88-2
Molecular Formula C14H20Cl2N4
Molecular Weight (g/mol) 315.242
MDL Number MFCD00078831
InChI Key PJBQYZZKGNOKNJ-UHFFFAOYSA-M
Synonym amprolium hydrochloride, amprolium hcl, amprolium, mepyrium, amprol, amprol 25, thiacoccid, amprol 25 per cent, amprolsol vet, corid vet
PubChem CID 8732
IUPAC Name 5-[(2-methylpyridin-1-ium-1-yl)methyl]-2-propylpyrimidin-4-amine;chloride;hydrochloride
SMILES CCCC1=NC=C(C(=N1)N)C[N+]2=CC=CC=C2C.Cl.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6478214 View Documents Alfa Aesar™
J6478214
25g Each for $103.95
 
AAJ6478222 View Documents Alfa Aesar™
J6478222
100g Each for $374.73
 
Specifications

Chemical Identifiers

137-88-2
315.242
PJBQYZZKGNOKNJ-UHFFFAOYSA-M
8732
CCCC1=NC=C(C(=N1)N)C[N+]2=CC=CC=C2C.Cl.[Cl-]
C14H20Cl2N4
MFCD00078831
amprolium hydrochloride, amprolium hcl, amprolium, mepyrium, amprol, amprol 25, thiacoccid, amprol 25 per cent, amprolsol vet, corid vet
5-[(2-methylpyridin-1-ium-1-yl)methyl]-2-propylpyrimidin-4-amine;chloride;hydrochloride

Specifications

White
137-88-2
MFCD00078831
amprolium hydrochloride, amprolium hcl, amprolium, mepyrium, amprol, amprol 25, thiacoccid, amprol 25 per cent, amprolsol vet, corid vet
CCCC1=NC=C(C(=N1)N)C[N+]2=CC=CC=C2C.Cl.[Cl-]
315.242
315.24
1-(4-Amino-2-n-propyl-5-pyrimidinylmethyl)-2-methylpyridinium chloride
25g
C14H20Cl2N4
4118699
PJBQYZZKGNOKNJ-UHFFFAOYSA-M
5-[(2-methylpyridin-1-ium-1-yl)methyl]-2-propylpyrimidin-4-amine;chloride;hydrochloride
8732
Powder
Safety and Handling

Safety and Handling

RTECSNumber : TJ565000