Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4,8-Tri-Boc-1,4,8,11-tetraazacyclotetradecane

$486.42 - $486.42

Chemical Identifiers

Chemical Name or Material 1,4,8-Tri-Boc-1,4,8,11-tetraazacyclotetradecane
CAS 170161-27-0
Molecular Formula C25H48N4O6
Formula Weight 500.68
MDL Number MFCD09263315
Synonym N,N',N''-Tri-Boc-cyclam; 1,4,8-Tris(tert-butoxycarbonyl)-1,4,8,11-tetraazacyclotetradecane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26741MD

 
alfa aesar™
H26741MD
250mg Each for $486.42
Description & Specifications

Specifications

Formula Weight 500.68
Chemical Name or Material 1,4,8-Tri-Boc-1,4,8,11-tetraazacyclotetradecane
Molecular Formula C25H48N4O6
MDL Number MFCD09263315
CAS 170161-27-0
Synonym N,N',N''-Tri-Boc-cyclam; 1,4,8-Tris(tert-butoxycarbonyl)-1,4,8,11-tetraazacyclotetradecane
Melting Point 48° to 54°C
Health Hazard 1 Warning
Quantity 250mg
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.