Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4,8,11-Tetrakis(ethoxycarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane

$96.97 - $241.66

Chemical Identifiers

Chemical Name or Material 1,4,8,11-Tetrakis(ethoxycarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane
CAS 126320-57-8
Molecular Formula C26H48N4O8
Formula Weight 544.69
MDL Number MFCD09263316
Synonym TETAEt; Cyclam-N,N',N'',N'''-tetraacetic acid tetraethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2675503

 
alfa aesar™
H2675503
1g Each for $241.66

AAH26755MD

 
alfa aesar™
H26755MD
250mg Each for $96.97
Description & Specifications

Specifications

Formula Weight 544.69
Chemical Name or Material 1,4,8,11-Tetrakis(ethoxycarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane
Molecular Formula C26H48N4O8
MDL Number MFCD09263316
CAS 126320-57-8
Synonym TETAEt; Cyclam-N,N',N'',N'''-tetraacetic acid tetraethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 87° to 89°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.