Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4,8,11-Tetrakis(dimethylaminocarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane

$357.03 - $357.03

Chemical Identifiers

Chemical Name or Material 1,4,8,11-Tetrakis(dimethylaminocarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane
CAS 345612-68-2
Molecular Formula C26H52N8O4
Formula Weight 540.75g/mol
MDL Number MFCD09263306
Synonym TETAMMe2; Cyclam-N,N',N'',N'''-tetrakis(N,N-dimethyl-2-acetamide)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2642203

 
alfa aesar™
H26422-03
1g Each for $357.03
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 1,4,8,11-Tetrakis(dimethylaminocarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane
CAS 345612-68-2
Molecular Formula C26H52N8O4
Formula Weight 540.75g/mol
MDL Number MFCD09263306
Synonym TETAMMe2; Cyclam-N,N',N'',N'''-tetrakis(N,N-dimethyl-2-acetamide)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.