Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4,8,11-Tetrakis(diethylaminocarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane

$167.26 - $424.91

Chemical Identifiers

Chemical Name or Material 1,4,8,11-Tetrakis(diethylaminocarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane
CAS 126320-56-7
Molecular Formula C34H68N8O4
Formula Weight 652.97
MDL Number MFCD09263304
Synonym TETAMEt2; Cyclam-N,N',N'',N'''-tetrakis(N,N-diethylacetamide)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH26367MD

  SDS
alfa aesar™
H26367MD
250mg Each for $167.26

AAH2636703

  SDS
alfa aesar™
H2636703
1g Each for $424.91
Description & Specifications

Specifications

Formula Weight 652.97
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 250mg
Synonym TETAMEt2; Cyclam-N,N',N'',N'''-tetrakis(N,N-diethylacetamide)
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 144° to 145°C
CAS 126320-56-7
MDL Number MFCD09263304
Chemical Name or Material 1,4,8,11-Tetrakis(diethylaminocarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane
Molecular Formula C34H68N8O4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.