missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4,8,11-Tetraazacyclotetradecane, 98+%

Manufacturer:  Alfa Aesar™ A1151606

 View more versions of this product

Catalog No. AAA1151606


 

Description

Description

Applications
1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications

Specifications

1,4,8,11-Tetraazacyclotetradecane
≥98%
MFCD00005105
Cyclam
C1CNCCNCCCNCCNC1
200.33
CHEBI:37401
184°C to 188°C
295-37-4
C10H24N4
111811
MDAXKAUIABOHTD-UHFFFAOYSA-N
1,4,8,11-tetrazacyclotetradecane
64964
200.33
5g

Chemical Identifiers

295-37-4
200.33
MDAXKAUIABOHTD-UHFFFAOYSA-N
64964
C10H24N4
MFCD00005105
Cyclam
CHEBI:37401
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XA5254800

SDS Documents