missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4,7-Tris(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane

$261.68 - $261.68

Chemical Identifiers

CAS 122555-91-3
Molecular Formula C26H50N4O6
Formula Weight 514.71
MDL Number MFCD05865261
Synonym DO3AtBu; Cyclen-N,N',N''-triacetic acid tri-tert-butyl ester
Chemical Name or Material 1,4,7-Tris(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH26736MD SDS Alfa Aesar™
H26736MD
250mg Each for $261.68
Specifications Description & Specifications

Chemical Identifiers

122555-91-3
514.71
DO3AtBu; Cyclen-N,N',N''-triacetic acid tri-tert-butyl ester
C26H50N4O6
MFCD05865261
1,4,7-Tris(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane

Specifications

122555-91-3
514.71
DO3AtBu; Cyclen-N,N',N''-triacetic acid tri-tert-butyl ester
1,4,7-Tris(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
C26H50N4O6
MFCD05865261
250mg

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning