missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4,7-Triazacyclononane trihydrochloride, 97%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 294315000

 View more versions of this product

Catalog No. AC294315000



Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chelation/Complexation reagent
Specifications

Specifications

1, 4, 7-Triazacyclononane trihydrochloride
58966-93-1
100.0
285.0°C to 290.0°C
Authentic
MFCD00074887
1,4,7-triazacyclononane tri hydrochloride, 1,4,7-triazacyclononane trihydrochloride, 1,4,7-triazacyclononanetrihydrochloride, 1,4,7-triazonane trihydrochloride, 1h-1,4,7-triazonine, octahydro-, trihydrochloride, acmc-1au73, c6h15n3.3hcl, ksc911s7j, octahydro-1h-1,4,7-triazonine trihydrochloride, pubchem21332
ITWBWJFEJCHKSN-UHFFFAOYSA-Q
1,4,7-triazonane-1,4,7-triium
2724990
Crystalline Powder
97%
96.0
White to Yellow
97%
C6H18N3
Glass bottle
500mg
C1C[NH2+]CC[NH2+]CC[NH2+]1
132.23
238.59
97%

Chemical Identifiers

58966-93-1
132.23
ITWBWJFEJCHKSN-UHFFFAOYSA-Q
2724990
C1C[NH2+]CC[NH2+]CC[NH2+]1
C6H18N3
MFCD00074887
1,4,7-triazacyclononane tri hydrochloride, 1,4,7-triazacyclononane trihydrochloride, 1,4,7-triazacyclononanetrihydrochloride, 1,4,7-triazonane trihydrochloride, 1h-1,4,7-triazonine, octahydro-, trihydrochloride, acmc-1au73, c6h15n3.3hcl, ksc911s7j, octahydro-1h-1,4,7-triazonine trihydrochloride, pubchem21332
1,4,7-triazonane-1,4,7-triium
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

TSCA : TSCA

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only