missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4,7-Triazacyclononane, TCI America™

Manufacturer:  TCI America T18781G

 View more versions of this product

Catalog No. T18781G



Specifications

Specifications

1,4,7-Triazacyclononane
White-Yellow
1g
C6H15N3
Octahydro-1H-1,4,7-triazonine
C1CNCCNCCN1
129.207
CHEBI:37405
Crystal-Powder at 20°C
113°C
45°C
4730-54-5
3259
ITWBWJFEJCHKSN-UHFFFAOYSA-N
1,4,7-triazonane
188318
129.21
≥98.0% (GC)

Chemical Identifiers

4730-54-5
129.207
Octahydro-1H-1,4,7-triazonine
CHEBI:37405
C1CNCCNCCN1
C6H15N3
ITWBWJFEJCHKSN-UHFFFAOYSA-N
188318
1,4,7-triazonane
Safety and Handling

Safety and Handling

TSCA : No

Documents