missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4,7,10-Tetrakis(ethoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane

$157.00 - $157.00

Chemical Identifiers

CAS 137076-50-7
Molecular Formula C24H44N4O8
Formula Weight 516.64
MDL Number MFCD09263313
Synonym DOTAEt; Cyclen-N,N',N'',N'''-tetraacetic acid tetraethyl ester
Chemical Name or Material 1,4,7,10-Tetrakis(ethoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH26646MD SDS Alfa Aesar™
H26646MD
250mg Each for $157.00
Specifications Description & Specifications

Chemical Identifiers

137076-50-7
516.64
DOTAEt; Cyclen-N,N',N'',N'''-tetraacetic acid tetraethyl ester
C24H44N4O8
MFCD09263313
1,4,7,10-Tetrakis(ethoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane

Specifications

137076-50-7
516.64
DOTAEt; Cyclen-N,N',N'',N'''-tetraacetic acid tetraethyl ester
1,4,7,10-Tetrakis(ethoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane
C24H44N4O8
MFCD09263313
250mg

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning